Bel-Art 183910001 Polycarbonate Lab Jack, 8" x 8" Top, 3" to 12" H, 15 Lb Capacity

Representative image only.

Bel-Art 183910001 Polycarbonate Lab Jack, 8" x 8" Top, 3" to 12" H, 15 Lb Capacity

Bel-Art | Mfr: 183910001| Item: 0805550

Autoclavable Max support weight is 15 lbs (6 kg)

$618.82CAD / Each

$618.82CAD / Each
Product Description

Key Features

  • Applications: Separation of peptides and proteins
  • 1 year warranty

More About this Item

This polycarbonate jack is perfect for use in highly corrosive atmospheres or for submersion in liquid baths—it won’t conduct electricity, corrode, or rust. Polyetherimide screw threads and bushings of PTFE withstand high temperatures during autoclaving.
Interchangeable parts are easily replaced in the event of breakage. The 8" x 8" top and bottom plates can be drilled for mounting to a wall. Use either top or bottom plate for support. (If more vertical stability is required, add weights to the bottom plate.)
Jack reaches a maximum height of 12"; closed height is 3". Support up to 15 lb.
Note: Crank jack up, then put object on top plate. Do not use jack to lift weight from bottom position. However weight may be cranked down.

1 Year Warranty
Specifications
  • DescriptionPolycarbonate Lab Jack, 8" x 8" Top, 3" to 12" H, 15 Lb Capacity
  • Warranty1 year
Reviews
Q&A