Bain Marie Container Snap-Tight, Positive-Lock Lid, HDPE, for 63530-16, -18
Representative image only.
Bain Marie Container Snap-Tight, Positive-Lock Lid, HDPE, for 63530-16, -18
| Item: 6353054
Positive lock for tight seal
$214.75CAD / Pkg of 6
BUY MORE & SAVE
  • 1-5$214.75 CAD / Pkg of 6
  • 6-29$193.28 CAD / Pkg of 6
  • 30-119$182.54 CAD / Pkg of 6
  • 120+$171.80 CAD / Pkg of 6
$214.75CAD / Pkg of 6
BUY MORE & SAVE
  • 1-5$214.75 CAD / Pkg of 6
  • 6-29$193.28 CAD / Pkg of 6
  • 30-119$182.54 CAD / Pkg of 6
  • 120+$171.80 CAD / Pkg of 6
Product Description
Key Features
  • NSF certified
More About this Item
Snap-tight, positive-lock lids are made of FDA compliant High-Density PolyEthylene (HDPE) and can be frozen, boiled, or chemically sterilized. Bain Marie HDPE containers are sold separately.
NSF international certified
Specifications
  • DescriptionBain Marie Container Snap-Tight, Positive-Lock Lid, HDPE, for 63530-16, -18