Aven Tools Tweezers, Stainless Steel, Straight, Ultra Fine Sharp Tips, 4-1/4"L
Representative image only.
Aven Tools Tweezers, Stainless Steel, Straight, Ultra Fine Sharp Tips, 4-1/4"L
Aven Tools | Mfr: 18056USA| Item: 0728706
Ideal for micro electronics and precision assembly
$18.86CAD / Pkg of 1
BUY MORE & SAVE
  • 1-5$18.86 CAD / Pkg of 1
  • 6-29$16.97 CAD / Pkg of 1
  • 30-119$16.03 CAD / Pkg of 1
  • 120+$15.09 CAD / Pkg of 1
$18.86CAD / Pkg of 1
BUY MORE & SAVE
  • 1-5$18.86 CAD / Pkg of 1
  • 6-29$16.97 CAD / Pkg of 1
  • 30-119$16.03 CAD / Pkg of 1
  • 120+$15.09 CAD / Pkg of 1
Product Description
Key Features
  • Lifetime warranty
  • Limited Lifetime Warranty
More About this Item
These finely crafted stainless steel tweezers feature hand finished points for accuracy and excellent tip hardness. Anti-glare finish reduces eye strain. The tweezers are also anti-magnetic, anti-acid, and acid resistant.
Limited Lifetime Warranty
Specifications
  • Overall Length (in)4 1/4
  • Overall Length (cm)10.78
  • MaterialStainless Steel
  • SterileNo
  • StyleStraight, Ultra Fine Sharp Tips
  • DescriptionTweezers, Stainless Steel, Straight, Ultra Fine Sharp Tips, 4-1/4"L
  • WarrantyLimited Lifetime