Aven Tools Tweezers, Stainless Steel, Serrated Straight Tips, 4.75" L
Representative image only.
Aven Tools Tweezers, Stainless Steel, Serrated Straight Tips, 4.75" L
Aven Tools | Mfr: 18037USA| Item: 0728703
Ideal for micro electronics and precision assembly
$21.81CAD / Pkg of 1
BUY MORE & SAVE
  • 1-5$21.81 CAD / Pkg of 1
  • 6-29$19.63 CAD / Pkg of 1
  • 30-119$18.54 CAD / Pkg of 1
  • 120+$17.45 CAD / Pkg of 1
$21.81CAD / Pkg of 1
BUY MORE & SAVE
  • 1-5$21.81 CAD / Pkg of 1
  • 6-29$19.63 CAD / Pkg of 1
  • 30-119$18.54 CAD / Pkg of 1
  • 120+$17.45 CAD / Pkg of 1
Product Description
Key Features
  • Lifetime warranty
More About this Item
These finely crafted stainless steel tweezers feature hand finished points for accuracy and excellent tip hardness. Anti-glare finish reduces eye strain. The tweezers are also anti-magnetic, anti-acid, and acid resistant.
Limited Lifetime Warranty
Specifications
  • Overall Length (in)4 3/4
  • Overall Length (cm)12.07
  • MaterialStainless Steel
  • SterileNo
  • StyleStraight, Thick, Flat, Strong Tips
  • DescriptionTweezers, Stainless Steel, Serrated Straight Tips, 4.75" L
  • WarrantyLimited Lifetime