Ashcroft 1105T Gauge Tool accessory Kit
Representative image only.
Ashcroft 1105T Gauge Tool accessory Kit
Ashcroft | Mfr: 1105T| Item: 0735970
$800.05CAD / Each
$800.05CAD / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Tool Kit can save time and money by servicing your own gauges. Kit includes everything you need for gauge maintenance: hand jack set (model 07359-60), screwdriver, five reamers, pin vise holder, wiggler, tweezers, and a soft carrying case that measures 9-3/8''L x 3-1/2''W x 1-3/4''H.
1 Year Warranty
Specifications
  • DescriptionGauge Tool accessory Kit