Arrow Mixing Products Model G-HD Medium/Heavy Duty, Gear-Driven Air Powered Mixer
Representative image only.
Arrow Mixing Products Model G-HD Medium/Heavy Duty, Gear-Driven Air Powered Mixer
Arrow Mixing Products | Mfr: G-HD| Item: 0468110
Safely use these air-drive mixers whenever electrical power is hazardous or impractical. Ideal for mixing solvents, lacquers, and volatile chemicals without the possibility of spark ignition.
$1,243.54CAD / Each
for an additional
$307.57CAD / Each
for an additional
$432.44CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$1,243.54CAD / Each
Buy It Together
Current Item
Arrow Mixing Products Model G-HD Medium/Heavy Duty, Gear-Driven Air Powered Mixer
Arrow Mixing Products Model G-HD Medium/Heavy Duty, Gear-Driven Air Powered Mixer
0468110
$1,243.54CAD / Each
+
Air lubricator, 1 to 20 scfm, 1/4" NPT(F)
0704270
$216.59CAD / Each
+
InnoCal NIST-Traceable Calibration; Mixer
1705003
$307.57CAD / Each
=
Total Price 1767.70 ADD 3 ITEMS TO CART
Product Description
Key Features
  • 1 year warranty
More About this Item
Explosion proof mixers are a no-brainer when working in hazardous environments. Distilleries, refineries, and many other plants often seek explosion proof mixers for their applications. The hazards they face are real, both physical and chemical in many cases. Mixers are also a common sight in these liquid processing facilities. Putting these together, these environments require mixers with hazardous duty ratings to prove they will not create an explosion hazard. Adding in the viscosity concerns and batch sizes common to most industrial processes, a high-torque gear-drive hazardous-duty gear-drive mixer is the most common and preferred solution to mixing needs. Choosing explosion proof mixers for industrial applications is one of the most difficult engineering challenges. It is part science and part art. Consideration must be made for: viscosity, specific gravity, temperature, speed, timing, material compatibility, particulates, tank type, size, and dimensions, mission, and compliance. Our experience, training, and relationships enable us to solve your industrial mixer challenge from the R&D bench to the production floor. If you have questions about explosion proof mixers contact an Application Specialist today.
Includes: 316 SS paddle assembly (3⁄8" dia x 12"L [10 mm dia x 305 mm L] shaft with 21⁄2" [64 mm] dia impeller), collet, 10"L x 9⁄16" dia (254 mm L x 14 mm dia) mounting rod, muffler, needle valve, and 4-ft (1.2-m) L air hose with fittings.
1 Year Warranty
Specifications
  • Material316 Stainless Steel
  • Min Speed (rpm)50
  • Max Speed (rpm)1200
  • Max Torque (in-oz)288
  • Motor hp1/3
  • DescriptionLow-cost, gear-drive air motor mixer with steel gears; 50-1200 rpm