Arrow Mixing Products Model 750 Heavy Duty, Direct-Drive Air Powered Mixer
Representative image only.
Arrow Mixing Products Model 750 Heavy Duty, Direct-Drive Air Powered Mixer
Arrow Mixing Products | Mfr: 750 W/18"SHFT/VPP-36/CPLG| Item: 0468615
$2,453.19CAD / Each
for an additional
$432.44CAD / Each
for an additional
$307.57CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$2,453.19CAD / Each
Buy It Together
Current Item
Arrow Mixing Products Model 750 Heavy Duty, Direct-Drive Air Powered Mixer
Arrow Mixing Products Model 750 Heavy Duty, Direct-Drive Air Powered Mixer
0468615
$2,453.19CAD / Each
+
Air lubricator, 1 to 20 scfm, 1/4" NPT(F)
0704270
$216.59CAD / Each
+
InnoCal NIST-Traceable Calibration; Mixer
1705003
$307.57CAD / Each
=
Total Price 2977.35 ADD 3 ITEMS TO CART
Product Description
Key Features
  • 1 year warranty
More About this Item
Safely use these air-drive mixers whenever electrical power is hazardous or impractical. Ideal for mixing solvents, lacquers, and volatile chemicals without the possibility of spark ignition.
Mixers accept an air input line pressure to 80 or 100 psi (10 to 12 cfm). A needle valve enables you to control mixing speed throughout the range for each mixer, and a built-in muffler ensures quiet operation. We recommend installing an air filter/regulator 07042-75 and an air lubricator 07042-70 in your air supply line.
Includes: 3⁄8" dia x 18"L (10 mm dia x 457 mm L), 316 SS shaft; 35⁄8" (92 mm) dia, 316 SS impeller; collet; 10"L x 5⁄8" dia (254 mm L x 16 mm dia) mounting rod; muffler; needle valve; and 4-ft (1.2-m) L air hose with fittings.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
1 Year Warranty
Specifications
  • Min Speed (rpm)300
  • Max Speed (rpm)3000
  • Max Torque (in-oz)448
  • Motor hp3/4
  • DescriptionLow-cost, Direct-drive Air Motor Mixer; 300-3,000 RPM