Ansell TouchNTuff® 92-616 Lightweight Disposable Nitrile Gloves, M (7, 5-8); 1500/CS (10 boxes of 150 gloves)
Representative image only.
Ansell TouchNTuff® 92-616 Lightweight Disposable Nitrile Gloves, M (7, 5-8); 1500/CS (10 boxes of 150 gloves)
Ansell | Mfr: 105132| Item: 8162070
Puncture and chemical splash resistant
$501.98CAD / Case of 1500
$501.98CAD / Case of 1500
Product Description
Key Features
  • Compliant with food handling requirements
  • Packaged 150 gloves per dispenser
  • Ambidextrous
More About this Item
These lightweight nitrile gloves provide you comfort and durability. Made of 100% nitrile to prevent Type 1 allergies. These gloves offer you puncture and chemical splash protection from hydrocarbons, fats, grease, and alcohols. Ideal for food processing applications. Packaged 150 gloves per dispenser to minimize packaging.Ten packages per case.
Specifications
  • Thickness (mil)3.1
  • MaterialNitrile
  • SizeMedium
  • Length (in)9 1/2
  • Length (cm)24
  • ColorBlue
  • GripTextured fingertips
  • CuffStandard
  • ComplianceFDA 21 CFR 177-2600
  • SterileNo
  • PowderedNo
  • DescriptionLightweight Disposable Nitrile Gloves, M (7, 5-8); 1500/CS (10 boxes of 150 gloves)