Ansell Microflex® V29 Blue Disposable Vinyl Gloves, Size L (8.5-9); 1000/CS (10 boxes of 100 gloves)
Representative image only.
Ansell Microflex® V29 Blue Disposable Vinyl Gloves, Size L (8.5-9); 1000/CS (10 boxes of 100 gloves)
Ansell | Mfr: 163310| Item: 8162079
Cost-effective protection for light duty tasks
$271.40CAD / Case of 1000
$271.40CAD / Case of 1000
Product Description
Key Features
  • Free from natural rubber latex
  • Produced without use of DEHP
  • Ambidextrous
More About this Item
These disposable blue vinyl gloves provide outstanding elasticity and tensile strength, ideal for food preparation, light-duty cleaning, and general maintenance. The gloves offer a smooth finish, giving you a more natural fit. Powder-free to reduce skin irritation. Disposable—for single use only.
Specifications
  • Thickness (mil)2.8
  • MaterialVinyl
  • SizeLarge
  • Length (in)9 1/2
  • Length (cm)24
  • ColorBlue
  • CuffStandard
  • ComplianceFDA 21 CFR 177-2600
  • SterileNo
  • PowderedNo
  • DescriptionBlue Disposable Vinyl Gloves, Size L (8.5-9); 1000/CS (10 boxes of 100 gloves)