Ansell Microflex® V28 Light Duty Disposable Vinyl Gloves, Size XL (9.5-10); 1000/CS (10 boxes of 100 gloves)
Representative image only.
Ansell Microflex® V28 Light Duty Disposable Vinyl Gloves, Size XL (9.5-10); 1000/CS (10 boxes of 100 gloves)
Ansell | Mfr: 163301| Item: 8162076
Cost-effective protection for light duty tasks
$178.13CAD / Case of 1000
$178.13CAD / Case of 1000
Product Description
Key Features
  • Free from natural rubber latex
  • Produced without use of DEHP
  • Ambidextrous
More About this Item
These disposable vinyl gloves are ideal for food handling, light-duty cleaning, and general-purpose use. The gloves offer a smooth finish, giving you a more natural fit. Powder-free to reduce skin irritation. Disposable—for single use only.
Specifications
  • MaterialVinyl
  • Thickness (mil)2.8
  • SizeX-Large
  • Length (in)9 1/2
  • Length (cm)24
  • ColorClear
  • CuffStandard
  • ComplianceFDA 21 CFR 177-2600
  • SterileNo
  • PowderedNo
  • DescriptionLight Duty Disposable Vinyl Gloves, Size XL (9.5-10.0)