Analytik Jena Pen-Ray 99000401 Power supply for 97606-08, 115 VAC/60 Hz

Representative image only.

Analytik Jena Pen-Ray 99000401 Power supply for 97606-08, 115 VAC/60 Hz

Analytik Jena | Mfr: 99000401| Item: 9760690

$1,135.98CAD / Each

$1,135.98CAD / Each

Buy It Together

Current Item

Analytik Jena Pen-Ray 99000401 Power supply for 97606-08, 115 VAC/60 Hz
Analytik Jena Pen-Ray 99000401 Power supply for 97606-08, 115 VAC/60 Hz
9760690

$1,135.98CAD / Each
+ + + + + +
Ultraviolet Crosslinker Replacement Tube; 302 nm
9760751

$106.23CAD / Each
=
Total Price 2965.19 ADD 7 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • 1 year warranty

More About this Item

Keep your ultraviolet equipment running and dependable with replacement bulbs and accessories.

1 Year Warranty
Specifications
  • Compatible With97606-08
  • Power (VAC)115
  • DescriptionPower supply for 97606-08, 115 VAC/60 Hz
  • Warranty1 year
Reviews
Q&A