AmScope™ High Precision Tweezer Set in Vinyl Pouch, 6 Piece Set
Representative image only.
AmScope™ High Precision Tweezer Set in Vinyl Pouch, 6 Piece Set
| Item: 7890510
Anti-acid – excellent corrosion resistance to most chemicals, salts, and acids
$81.04CAD / Each
$81.04CAD / Each
Product Description
Includes
5" Straight Precise Tip Tweezer Model AA, 4-1/2" Straight Flat Edge Tweezer Model OO, 4-3/4" Straight Flat Edge Tweezer Model OOD, 4-1/4" Straight Fine Point Tweezer Model 3c, 4-1/4" Tapered Ultra Fine Point Tweezer Model 5, and 4-3/4" Curved Fine Tip Tweezer Model 7
Key Features
  • Manufactured from 300 series anti-magnetic stainless steel
  • Resistant to phosphoric acid at most concentrations, sulfuric acid at concentrations below 10%, hydrochloric acid, acetic acid, and citric acid
  • 1 year warranty
More About this Item
Primarily used for microscopy applications, these tweezers are engineered to meet and exceed the requirements for electronics assembly, labs, clean rooms and circuit board repair and rework. These high precision metal tweezers are made of the highest quality materials and have tips that are perfectly balanced. They offer tip symmetry for precision work.
1 Year Warranty
Specifications
  • DescriptionHigh Precision Tweezers, 6 Piece Set in Vinyl Pouch