Amprobe VP-440 Non-Contact VoltProbe
Representative image only.
Amprobe VP-440 Non-Contact VoltProbe
Amprobe | Mfr: VP-440| Item: 2000892
$81.94CAD / Each
for an additional
$195.59CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$81.94CAD / Each
Buy It Together
Current Item
Amprobe VP-440 Non-Contact VoltProbe
Amprobe VP-440 Non-Contact VoltProbe
2000892
$81.94CAD / Each
+ =
Total Price 277.53 ADD 2 ITEMS TO CART
Product Description
Key Features
  • 1 year warranty
More About this Item
  • Detect AC voltage (100-600V)
  • Use non-contact method of voltage testing
  • Totally insulated, no exposed metal or conductive parts
  • Check for AC voltages without necessity of disconnecting the electrical system
  • Test for AC voltage cables, circuit breakers, wall sockets, junction boxes, fuses, etc.
  • CAT III 600V
1 Year Warranty
Specifications
  • DC Max (Volts)600
  • AC Max (Volts)600
  • Safety RatingCAT III 600V, IEC 1010-1
  • DescriptionNon-Contact VoltProbe