Amprobe TMD-10 Thermocouple Thermometer, Type J/K
Representative image only.
Amprobe TMD-10 Thermocouple Thermometer, Type J/K
Amprobe | Mfr: TMD-10| Item: 9600103
Compact and rugged design featuring a large display
$386.97CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$386.97CAD / Each
Buy It Together
Current Item
Amprobe TMD-10 Thermocouple Thermometer, Type J/K
Amprobe TMD-10 Thermocouple Thermometer, Type J/K
9600103
$386.97CAD / Each
+ + =
Total Price 978.53 ADD 3 ITEMS TO CART
Product Description
Includes
2 type-k probes, carrying case, and 1 9V battery.
Key Features
  • Accepts any type K or J thermocouple probe with miniconnector
  • Dual temperature for easy Super-Heat and Sub-Cool calculations
  • Hold function to retain reading on the screen for later viewing
  • Relative reading (T1 – T2)
  • Selectable deg F or deg C
  • Min/Max recording
  • Low battery indication
  • 1 year warranty
More About this Item
The TMD-10 is a dual input thermometer that accepts type K and J thermocouples and is ideal for HVAC/R technicians, electricians, research, industrial process, and quality control. The dual inputs allow easy superheat and sub-cool temperature measurement as well as cut-out and in temperature measurements. The triple display shows T1, T2, and T1 - T2 and includes Hold, Min/Max, Relative, and Selectable deg F or deg C.
1 Year Warranty
Specifications
  • Meter Measurement TypeThermocouple
  • Number Of Probe Channels2
  • Types Of Probes AcceptedType J, Type K
  • Min Temperature (° F)-328
  • Max Temperature (° F)2498
  • Min Temperature (° C)-200
  • Max Temperature (° C)1370
  • Accuracy±0.05% of reading +1.4°F (+0.7°C)
  • Resolution0.1°
  • Low Battery IndicatorYes
  • Display Type4.5-digit LCD
  • No. of Probe(s) Included1
  • Battery1 9V (included)
  • DescriptionThermocouple Thermometer, Type J/K