Amprobe 5XP Compact DMM with non-contact voltage detector
Representative image only.
Amprobe 5XP Compact DMM with non-contact voltage detector
Amprobe | Mfr: 5XP-A| Item: 2000769
Available for sale only in the United States and Canada
$210.93CAD / Each
for an additional
$195.59CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$210.93CAD / Each
Buy It Together
Current Item
Amprobe 5XP Compact DMM with non-contact voltage detector
Amprobe 5XP Compact DMM with non-contact voltage detector
2000769
$210.93CAD / Each
+ =
Total Price 406.52 ADD 2 ITEMS TO CART
Product Description
Key Features
  • 1 year warranty
More About this Item
Each Meterman multimeter has unique extras such as Voltect, Non-Contact Voltage (NCV) detection which allows the quick detection of dangerous AC voltages without probing, plus the exclusive Magne-Trip holster with magnetic hanging strap that frees both hands for your work.
All three models include continuity and diode test features. These multimeters are safety rated: CATI 1000V / CAT II 600V / CAT III 300V CE 1010/US-CSA-C. Each DMM ships complete with battery, test probes, Magne-Grip holster and one year warranty. The 35XP ships with a temperature probe.
1 Year Warranty
Specifications
  • Resistance Min (Ω)200
  • Resistance Max (MΩ)20
  • DisplayManual Ranging, 2000 ct
  • Safety RatingCAT II 600V, CAT III 300V
  • DescriptionCompact DMM with non-contact voltage detector