Allsafe 3010343 Shield, Blue Frame, Clear Lens
Representative image only.
Allsafe 3010343 Shield, Blue Frame, Clear Lens
Allsafe | Mfr: 18629| Item: 8165718
Greater range of visibility with a more comfortable fit
$87.96CAD / Each
BUY MORE & SAVE
  • 1-9$87.96 CAD / Each
  • 10+$79.16 CAD / Each
$87.96CAD / Each
BUY MORE & SAVE
  • 1-9$87.96 CAD / Each
  • 10+$79.16 CAD / Each
Buy It Together
Current Item
Allsafe 3010343 Shield, Blue Frame, Clear Lens
Allsafe 3010343 Shield, Blue Frame, Clear Lens
8165718
$87.96CAD / Each
+ =
Total Price 87.96 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 1 year warranty
More About this Item
Lightweight goggle provides superior impact and chemical splash protection. Cylindrical-shaped lens offers greater range of visibility. The 0.060"-thick polycarbonate lens features antifog coating and provides 99.9% UV protection. The soft frame is pliable and conforms to worker’s face for a comfortable fit.
Compliance: ANSI Z87.1+ (High impact)
Model 81657-18 features a detachable curved face shield for full-face protection.
1 Year Warranty
Specifications
  • Frame ColorBlue
  • Lens ColorClear
  • ComplianceANSI Z87.1+ (High Impact)
  • DescriptionShield, Blue Frame, Clear Lens