AEMC 407 Clamp Meter, 1000 AC, 1500 A DC with Bluetooth capability

Representative image only.

AEMC 407 Clamp Meter, 1000 AC, 1500 A DC with Bluetooth capability

AEMC | Mfr: 2139.51| Item: 2004336

Models with Bluetooth communication capability provide additional flexibility

$978.40CAD / Each

for an additional
$195.59CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$978.40CAD / Each

Buy It Together

Current Item

AEMC 407 Clamp Meter, 1000 AC, 1500 A DC with Bluetooth capability
AEMC 407 Clamp Meter, 1000 AC, 1500 A DC with Bluetooth capability
2004336

$978.40CAD / Each
+ =
Total Price 1173.99 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Measure’s up to 1000 V AC, DC and AC+DC with resolution to 10 mV
  • Measures single and three phase power (Real, Reactive and Apparent) up to 3 MW with resolution to 1W
  • Measures voltage frequency to 20 kHz with 0.1 Hz resolution
  • Auto selects AC or DC measurement
  • Measures Harmonics up to the 25th
  • Records up to 1000 measurements
  • Bluetooth communication and DataView® software compatible (models 407, and 607)
  • 1 year warranty
1 Year WarrantyCanadian Standards AssociationIP54 RatedUL Listed.
Specifications
  • DC Max (Volts)1000
  • DC Max (amps)1500
  • Current (AC/DC)1000 A
  • AC Max (Volts)1000
  • AC Max (amps)1000
  • Resistance Max (KΩ)100
  • ContinuityYes (<40 Ω)
  • Count1000
  • Current Frequency2 kHz
  • Jaw Size1.89
  • Safety RatingCAT IV 1000 V
  • PowerFour AA batteries
  • Width (in)3.625
  • Height (in)11
  • Depth (in)1.625
  • DescriptionClamp Meter, 1000 AC, 1500 A DC with Bluetooth capability
  • Warranty1 Year
Reviews
Q&A