Advantec 08010010 Residual Chlorine Test Strips, 10 to 50 ppm; 300 Strips/Pk

Representative image only.

Advantec 08010010 Residual Chlorine Test Strips, 10 to 50 ppm; 300 Strips/Pk

Advantec | Mfr: 08010010| Item: 5920123

Ideal for rapid check of residual chlorine levels in water samples

$79.12CAD / Pkg of 1

$79.12CAD / Pkg of 1
Product Description

Key Features

  • Detects chlorine as hypochlorus acid (HClO) or the hypochlorite ion (ClO-)
  • Chemically-treated paper strips change color and can be easily used in the lab or out in the field

More About this Item

These test strips are perfect for quickly estimating residual chlorine levels as hypochlorus acid (HClO) and hypochlorite ions (ClO-) in samples treated with sodium hypochlorite (NaClO). Easy to hold strips are 1/4" W x 1.5" L (7 x 40 mm) — simply dip a strip into your sample, wait 60 seconds, and compare the color change of the strip to the chart on the bottle.

Specifications
  • TestChlorine
  • Measurement Points10 to 50 ppm
  • Reagent PlatformStrips
  • Tests Per Kit300
  • DescriptionResidual Chlorine Test Strips, 10 to 50 ppm; 300 Strips/Pk
Reviews
Q&A