Additel 9080 Cold Junction Compensation Kit for Process Calibrators
Representative image only.
Additel 9080 Cold Junction Compensation Kit for Process Calibrators
Additel | Mfr: 9080| Item: 1610683
A wide array of Additel accessories to support your Additel instruments
$844.25CAD / Each
$844.25CAD / Each
Product Description
More About this Item
Choose from a wide range of accessories for your Additel meters including replacement batteries, software, and carrying cases.
Includes thermocouple plug and compensation cable. Compatible with type S, R, B, K, J, T, E, and N thermocouples.
Specifications
  • DescriptionCold Junction Compensation Kit for Process Calibrators