Adam Equipment 3014011014 RS-232 Interface Cable for AE 503 and AE 504 Indicator
Representative image only.
Adam Equipment 3014011014 RS-232 Interface Cable for AE 503 and AE 504 Indicator
Adam Equipment | Mfr: 3014011014| Item: 1111954
Quick communication for your Adam Equipment balances
$58.94CAD / Each
$58.94CAD / Each
Product Description
Key Features
  • Streamline data collection
  • No longer need to input readings manually
  • 1 year warranty
More About this Item
Choose from a wide range of communication cables for your Adam Equiptment balance.
Use this male to female RS-232 interface cable to connect the balance or weighing scale to other devices such as a ATP thermal printer or computer.
1 Year Warranty
Specifications
  • Accessory TypeRS-232 cable
  • Connector TypeRS-232
  • RS-232 InterfaceYES
  • DescriptionRS-232 Interface Cable for AE 503 and AE 504 Indicator