AccuMac AM8060 Dual-Channel Precision Benchtop Thermometer
Representative image only.
AccuMac AM8060 Dual-Channel Precision Benchtop Thermometer
AccuMac | Mfr: AM8060| Item: 9045271
High accuracy, fast readings and great stability
$5,541.42CAD / Each
$5,541.42CAD / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
AccuMac feature high accuracy, fast readings and great stability.Aailable is dual and single channels models. The dual channelallow measurements and displays of two PRTs simultaneously. All of the readings can be stored in a USB flash drive or transferred to PC through an USB cable.
Both Precision Thermometers allows users to choose ITS-90, IEC-751 (DIN), or Callendar-Van Dusen conversion methods to response to various PRTs. Users can also choose to key in calibration data of each PRT to ensure the best accuracy. They are outstanding choices for precision temperature measurements and calibrations using PRTs, Drywell Calibrators and Temperature Bath.
1 Year Warranty
Specifications
  • Probe TypePRT/RTD
  • Min Temperature (° F)-328
  • Max Temperature (° F)1562
  • Min Temperature (° C)-200
  • Max Temperature (° C)850
  • DescriptionDual-Channel Precision Benchtop Thermometer