Accuform ZFD889VP NFPA Hazardous Materials Classification, Sign Kit, 14" X 10", plastic

Representative image only.

Accuform ZFD889VP NFPA Hazardous Materials Classification, Sign Kit, 14" X 10", plastic

Accuform | Mfr: ZFD889VP| Item: 4101004

Signs and kit to help you with regulatory facility compliance

$53.19CAD / Each

$53.19CAD / Each
Product Description

More About this Item

These signs display the four-color National Fire protection Association (NFPA) diamond format. Signs feature blank hazard rating panels — write in the information for your specific use. Each sign measures 14" x 10" (35.6 x 25.4 cm).

Kit contains one sign, NFPA wallet guide, and adhesive numbers and hazard symbols which are printed black on clear adhesive polyester to fit the hazard panels.

Specifications
  • HeaderHAZARDOUS
  • LanguagesEnglish
  • MaterialPlastic
  • Width (in)10
  • Height (in)14
  • DescriptionNFPA Hazardous Materials Classification, Sign Kit, 14" X 10", plastic
Reviews
Q&A