Accuform ZFD600XT NFPA Placard, Sign Only, 6" X 6", plastic

Representative image only.

Accuform ZFD600XT NFPA Placard, Sign Only, 6" X 6", plastic

Accuform | Mfr: ZFD600XT| Item: 4101021

For on-the-spot classification of potentially hazardous chemicals

$19.89CAD / Each

$19.89CAD / Each
Product Description

More About this Item

These National Fire Protection Association (NFPA) Placards offer easy of installation and convenience. Blank placards are printed with NFPA color coding; simply write in the appropriate number rating in each quadrant.

Specifications
  • HeaderNFPA PLACARD
  • LanguagesEnglish
  • MaterialPlastic
  • Width (in)6
  • Height (in)6
  • DescriptionNFPA Placard, Sign Only, 6" X 6", plastic
Reviews
Q&A