Accuform ZFD101EV NFPA Placard, Sign Kit, 10" X 10", adhesive vinyl

Representative image only.

Accuform ZFD101EV NFPA Placard, Sign Kit, 10" X 10", adhesive vinyl

Accuform | Mfr: ZFD101EV| Item: 4101022

For on-the-spot classification of potentially hazardous chemicals

$68.22CAD / Each

$68.22CAD / Each
Product Description

More About this Item

These National Fire Protection Association (NFPA) Placards offer easy of installation and convenience. Blank placards are printed with NFPA color coding; simply write in the appropriate number rating in each quadrant.

Kit includes one placard, NFPA wallet guide, and numbers and symbols preprinted on clear adhesive polyester hazard panels.

Specifications
  • HeaderNFPA PLACARD
  • LanguagesEnglish
  • MaterialAdhesive Vinyl
  • Width (in)10
  • Height (in)10
  • DescriptionNFPA Placard, Sign Kit, 10" X 10", adhesive vinyl
Reviews
Q&A