Accuform MRDM102VS ANSI Sign,Danger-Do Not Enter Authorized Only, 7" x 10",Adhesive Vinyl

Representative image only.

Accuform MRDM102VS ANSI Sign,Danger-Do Not Enter Authorized Only, 7" x 10",Adhesive Vinyl

Accuform | Mfr: MRDM102VS| Item: 5071206

Use this sign to prevent unauthorized entry

$15.03CAD / Each

$15.03CAD / Each
Product Description

More About this Item

ANSI-style signs include a visual alert symbol (triangle with an exclamation point), a large symbol, and a hazard message. The large symbol gives a quick and easily understand indication of the potential hazard. Recognized by OSHA as an acceptable safety notification device.

ANSI Danger Signs (Red) indicate a hazardous situation which, if not avoided, will result in death or serious injury.

Specifications
  • HeaderDANGER
  • LanguagesEnglish
  • MaterialVinyl
  • Width (in)10
  • Height (in)7
  • DescriptionANSI Sign,Danger-Do Not Enter Authorized Only, 7" x 10",Adhesive Vinyl
Reviews
Q&A