Accuform MPPE791VP Safety Sign, Caution - Eye Protection Required, 7" X 10", Plastic

Representative image only.

Accuform MPPE791VP Safety Sign, Caution - Eye Protection Required, 7" X 10", Plastic

Accuform | Mfr: MPPE791VP| Item: 5071060

$17.68CAD / Each

$17.68CAD / Each
Product Description

More About this Item

These signs are intended to indicate potential, type, and degree of the hazard that may lead to accidental injury or property damage. Signs define specific hazards by conveying a clear, easily understood message. The sign headers and colors used in accident prevention safety signs are specified by OSHA and ANSI to designate the severity of the hazard. Each color has a specific role.

CAUTION (yellow): Indicates a potentially hazardous situation which, if not avoided, may result in minor or moderate injury.

Plastic is 0.055” polyethylene plastic and is good for medium duty indoor environments and sheltered outdoor environments.

Specifications
  • LegendEYE PROTECTION REQUIRED
  • HeaderCAUTION
  • LanguagesEnglish
  • MaterialPlastic
  • Width (in)10
  • Height (in)7
  • DescriptionSafety Sign, Caution - Eye Protection Required, 7" X 10", Plastic
Reviews
Q&A