Accuform MPPE791VA Safety Sign, Caution - Eye Protection Required, 7" X 10", Aluminum

Representative image only.

Accuform MPPE791VA Safety Sign, Caution - Eye Protection Required, 7" X 10", Aluminum

Accuform | Mfr: MPPE791VA| Item: 5071061

$27.55CAD / Each

$27.55CAD / Each
Product Description

Key Features

  • 5 year warranty

More About this Item

These signs are intended to indicate potential, type, and degree of the hazard that may lead to accidental injury or property damage. Signs define specific hazards by conveying a clear, easily understood message. The sign headers and colors used in accident prevention safety signs are specified by OSHA and ANSI to designate the severity of the hazard. Each color has a specific role.

CAUTION (yellow): Indicates a potentially hazardous situation which, if not avoided, may result in minor or moderate injury.

Aluminum is 0.040” thick with overlaminate coat for long-term outdoor exposure. It withstands splashes and washdown without rusting.

5 Year Warranty
Specifications
  • LegendEYE PROTECTION REQUIRED
  • HeaderCAUTION
  • LanguagesEnglish
  • MaterialAluminum
  • Width (in)10
  • Height (in)7
  • DescriptionSafety Sign, Caution - Eye Protection Required, 7" X 10", Aluminum
  • Warranty5 years
Reviews
Q&A