Accuform MFXG423VS Safety Sign, Fire Extinguisher (symbol), 10" x 7", Adhesive Vinyl

Representative image only.

Accuform MFXG423VS Safety Sign, Fire Extinguisher (symbol), 10" x 7", Adhesive Vinyl

Accuform | Mfr: MFXG423VS| Item: 6071055

$15.03CAD / Each

$15.03CAD / Each
Product Description

More About this Item

These signs are intended to indicate potential, type, and degree of the hazard that may lead to accidental injury or property damage. They are intended to define specific hazards by conveying a clear, easily understood message. The sign headers and colors used in accident prevention safety signs are specified by OSHA and ANSI to designate the severity of the hazard. Each color has a specific role.

DANGER (red): Indicates an imminently hazardous situation which, if not avoided, will result in death or serious injury.

Specifications
  • HeaderFIRE EXTINGUISHER
  • LanguagesEnglish
  • MaterialVinyl
  • Width (in)10
  • Height (in)7
  • DescriptionSafety Sign, Fire Extinguisher (symbol), 10" x 7", Adhesive Vinyl
Reviews
Q&A