Accuform MCHL252VP ANSI Sign, Danger-Hot, 10" x 14", Plastic

Representative image only.

Accuform MCHL252VP ANSI Sign, Danger-Hot, 10" x 14", Plastic

Accuform | Mfr: MCHL252VP| Item: 5071234

Protect your workers by clearly displaying possible hot hazards

$25.05CAD / Each

$25.05CAD / Each
Product Description

More About this Item

ANSI-style signs include a visual alert symbol (triangle with an exclamation point), a large symbol, and a hazard message. The large symbol gives a quick and easily understand indication of the potential hazard. Recognized by OSHA as an acceptable safety notification device.

ANSI Danger Signs (Red) indicate a hazardous situation which, if not avoided, will result in death or serious injury.

Specifications
  • HeaderDANGER
  • LanguagesEnglish
  • MaterialPlastic
  • Width (in)14
  • Height (in)10
  • DescriptionANSI Sign, Danger-Hot, 10" x 14", Plastic
Reviews
Q&A