Accuform MCHL127VS Safety Sign, Danger - Hot (with icon), 7" x 10", Adhesive Vinyl

Representative image only.

Accuform MCHL127VS Safety Sign, Danger - Hot (with icon), 7" x 10", Adhesive Vinyl

Accuform | Mfr: MCHL127VS| Item: 4071181

Protect your workers by clearly displaying possible hot areas

$15.03CAD / Each

$15.03CAD / Each
Product Description

More About this Item

These signs are intended to indicate potential, type, and degree of the hazard that may lead to accidental injury or property damage. Signs define specific hazards by conveying a clear, easily understood message. The sign headers and colors used in accident prevention safety signs are specified by OSHA and ANSI to designate the severity of the hazard. Each color has a specific role.
DANGER (red): indicates an imminently hazardous situation which, if not avoided, will result in death or serious injury.

Specifications
  • HeaderDANGER
  • LanguagesEnglish
  • MaterialVinyl
  • Width (in)10
  • Height (in)7
  • DescriptionSafety Sign, Danger - Hot (with icon), 7" x 10", Adhesive Vinyl
Reviews
Q&A