Accuform MBLEV6 NFPA Placard, Sign Only, 6" X 6", adhesive vinyl

Representative image only.

Accuform MBLEV6 NFPA Placard, Sign Only, 6" X 6", adhesive vinyl

Accuform | Mfr: MBLEV6| Item: 4101020

For on-the-spot classification of potentially hazardous chemicals

$11.20CAD / Each

$11.20CAD / Each
Product Description

More About this Item

These National Fire Protection Association (NFPA) Placards offer easy of installation and convenience. Blank placards are printed with NFPA color coding; simply write in the appropriate number rating in each quadrant.

Specifications
  • HeaderNFPA PLACARD
  • LanguagesEnglish
  • MaterialAdhesive Vinyl
  • Width (in)6
  • Height (in)6
  • DescriptionNFPA Placard, Sign Only, 6" X 6", adhesive vinyl
Reviews
Q&A