Accuform MADM486VS Safety Sign, No Exit, 7" X 10", Adhesive Vinyl

Representative image only.

Accuform MADM486VS Safety Sign, No Exit, 7" X 10", Adhesive Vinyl

Accuform | Mfr: MADM486VS| Item: 6071046

$15.03CAD / Each

$15.03CAD / Each
Product Description

More About this Item

These signs are intended to indicate potential, type, and degree of the hazard that may lead to accidental injury or property damage. They are intended to define specific hazards by conveying a clear, easily understood message. The sign headers and colors used in accident prevention safety signs are specified by OSHA and ANSI to designate the severity of the hazard. Each color has a specific role.

SAFETY FIRST
Indicates general instructions relative to safe work practices, remind of proper safety procedures, and indicate the location of safety equipment.

Signs are available in Adhesive Vinyl (VS), Plastic (VP), and Aluminum (VA) to best suit your application. VS is a flexible 4-mil pressure sensitive vinyl with adhesive back and peel-off liner. This material is ideal for low cost, lig

Specifications
  • HeaderEXIT
  • LanguagesEnglish
  • MaterialVinyl
  • Width (in)10
  • Height (in)7
  • DescriptionSafety Sign, No Exit, 7" X 10", Adhesive Vinyl
Reviews
Q&A