Accuform LZH614EV2 GHS Label, Corrosion Pictogram, 2" x 2", adhesive poly, 250/roll

Representative image only.

Accuform LZH614EV2 GHS Label, Corrosion Pictogram, 2" x 2", adhesive poly, 250/roll

Accuform | Mfr: LZH614EV2| Item: 5071278

$52.01CAD / Roll of 250

$52.01CAD / Roll of 250
Product Description

More About this Item

GHS (Globally Harmonized System) is a worldwide program for classifying chemicals in the workplace. GHS recognizes nine hazard pictograms. The pictograms identify the hazard which helps users take proper precautions to minimize adverse effects.
GHS Corrosion pictogram indicates the following potential hazard: skin corrosion/burns, eye damage, and corrosive to metals.

Specifications
  • HeaderCORROSION
  • LanguagesEnglish
  • MaterialAdhesive Poly
  • Width (in)2
  • Height (in)2
  • Quantity250/roll
  • DescriptionGHS Label, Corrosion Pictogram, 2" x 2", adhesive poly, 250/roll
Reviews
Q&A