Accuform LZH611PS2 GHS Label, Flame Pictogram, 2" x 2", adhesive coated paper, 250/roll

Representative image only.

Accuform LZH611PS2 GHS Label, Flame Pictogram, 2" x 2", adhesive coated paper, 250/roll

Accuform | Mfr: LZH611PS2| Item: 5071252

$33.59CAD / Roll of 250

$33.59CAD / Roll of 250
Product Description

More About this Item

GHS (Globally Harmonized System) is a worldwide program for classifying chemicals in the workplace. GHS recognizes nine hazard pictograms. The pictograms identify the hazard which helps users take proper precautions to minimize adverse effects.
GHS Flame pictogram indicates the following potential hazards: flammables, pyrophorics, self-heating, emits flammable gas, self-reactives, and organic peroxides.

Specifications
  • HeaderFLAME
  • LanguagesEnglish
  • MaterialAdhesive Paper
  • Width (in)2
  • Height (in)2
  • Quantity250/roll
  • DescriptionGHS Label, Flame Pictogram, 2" x 2", adhesive coated paper, 250/roll
Reviews
Q&A