Accuform LZH609PS5 GHS Label, Health Hazard Pictogram, 1" x 1", adhesive paper, 500/roll

Representative image only.

Accuform LZH609PS5 GHS Label, Health Hazard Pictogram, 1" x 1", adhesive paper, 500/roll

Accuform | Mfr: LZH609PS5| Item: 5071317

$46.26CAD / Roll of 500

$46.26CAD / Roll of 500
Product Description

More About this Item

GHS (Globally Harmonized System) is a worldwide program for classifying chemicals in the workplace. GHS recognizes nine hazard pictograms. The pictograms identify the hazard which helps users take proper precautions to minimize adverse effects.
GHS Health Hazard pictogram indicates the following potential hazard: carcinogen, mutagenicity, reproductive toxicity, respiratory sensitizer, target organ toxicity, aspiration toxicity.

Specifications
  • HeaderHEALTH HAZARD
  • LanguagesEnglish
  • MaterialAdhesive Paper
  • Width (in)1
  • Height (in)1
  • Quantity500/roll
  • DescriptionGHS Label, Health Hazard Pictogram, 1" x 1", adhesive paper, 500/roll
Reviews
Q&A