Accuform LZH607PS5 GHS Label,Exclamation Point Pictogram,1" x 1",adhesive paper,500/roll

Representative image only.

Accuform LZH607PS5 GHS Label,Exclamation Point Pictogram,1" x 1",adhesive paper,500/roll

Accuform | Mfr: LZH607PS5| Item: 5071301

$46.26CAD / Roll of 500

$46.26CAD / Roll of 500
Product Description

More About this Item

GHS (Globally Harmonized System) is a worldwide program for classifying chemicals in the workplace. GHS recognizes nine hazard pictograms. The pictograms identify the hazard which helps users take proper precautions to minimize adverse effects.
GHS Exclamation Point pictogram indicates the following potential hazard: irritant (skin and eye), skin sensitizer, acute toxicity, narcotic effects, respiratory tract irritant, hazardous to ozone layer.

Specifications
  • HeaderEXCLAMATION POINT
  • LanguagesEnglish
  • MaterialAdhesive Paper
  • Width (in)1
  • Height (in)1
  • Quantity500/roll
  • DescriptionGHS Label,Exclamation Point Pictogram,1" x 1",adhesive paper,500/roll
Reviews
Q&A