Accuform LZH606PS2 GHS Label,Skull/Crossbones Pictogram,1" x 1",adhesive paper,250/roll

Representative image only.

Accuform LZH606PS2 GHS Label,Skull/Crossbones Pictogram,1" x 1",adhesive paper,250/roll

Accuform | Mfr: LZH606PS2| Item: 5071288

$28.44CAD / Roll of 250

$28.44CAD / Roll of 250
Product Description

More About this Item

GHS (Globally Harmonized System) is a worldwide program for classifying chemicals in the workplace. GHS recognizes nine hazard pictograms. The pictograms identify the hazard which helps users take proper precautions to minimize adverse effects.
GHS Skull and Crossbones pictogram indicates the following potential hazard: acute toxicity (fatal or toxic).

Specifications
  • HeaderSKULL/CROSSBONES
  • LanguagesEnglish
  • MaterialAdhesive Paper
  • Width (in)1
  • Height (in)1
  • Quantity250/roll
  • DescriptionGHS Label,Skull/Crossbones Pictogram,1" x 1",adhesive paper,250/roll
Reviews
Q&A