Accuform LZH604PS5 GHS Label, Corrosion Pictogram, 1" x 1", adhesive paper, 500/roll

Representative image only.

Accuform LZH604PS5 GHS Label, Corrosion Pictogram, 1" x 1", adhesive paper, 500/roll

Accuform | Mfr: LZH604PS5| Item: 5071273

$46.26CAD / Roll of 500

$46.26CAD / Roll of 500
Product Description

More About this Item

GHS (Globally Harmonized System) is a worldwide program for classifying chemicals in the workplace. GHS recognizes nine hazard pictograms. The pictograms identify the hazard which helps users take proper precautions to minimize adverse effects.
GHS Corrosion pictogram indicates the following potential hazard: skin corrosion/burns, eye damage, and corrosive to metals.

Specifications
  • HeaderCORROSION
  • LanguagesEnglish
  • MaterialAdhesive Paper
  • Width (in)1
  • Height (in)1
  • Quantity500/roll
  • DescriptionGHS Label, Corrosion Pictogram, 1" x 1", adhesive paper, 500/roll
Reviews
Q&A