Accuform LZH603EV2 GHS Label,Exploding Bomb Pictogram,1" x 1",adhesive poly,250/roll

Representative image only.

Accuform LZH603EV2 GHS Label,Exploding Bomb Pictogram,1" x 1",adhesive poly,250/roll

Accuform | Mfr: LZH603EV2| Item: 5071266

$43.91CAD / Roll of 250

$43.91CAD / Roll of 250
Product Description

More About this Item

GHS (Globally Harmonized System) is a worldwide program for classifying chemicals in the workplace. GHS recognizes nine hazard pictograms. The pictograms identify the hazard which helps users take proper precautions to minimize adverse effects.
GHS Exploding Bomb pictogram indicates the following potential hazard: explosives, self-reactives, and organic peroxides.

Specifications
  • HeaderEXPLODING BOMB
  • LanguagesEnglish
  • MaterialAdhesive Poly
  • Width (in)1
  • Height (in)1
  • Quantity250/roll
  • DescriptionGHS Label,Exploding Bomb Pictogram,1" x 1",adhesive poly,250/roll
Reviews
Q&A