Accuform LZH602PS2 GHS Label,Flame Over Circle Pictogram,1" x 1",adhesive paper,250/roll

Representative image only.

Accuform LZH602PS2 GHS Label,Flame Over Circle Pictogram,1" x 1",adhesive paper,250/roll

Accuform | Mfr: LZH602PS2| Item: 5071256

$28.44CAD / Roll of 250

$28.44CAD / Roll of 250
Product Description

More About this Item

GHS (Globally Harmonized System) is a worldwide program for classifying chemicals in the workplace. GHS recognizes nine hazard pictograms. The pictograms identify the hazard which helps users take proper precautions to minimize adverse effects.
GHS Flame Over Circle pictogram indicates the following potential hazard: oxidizers

Specifications
  • HeaderFLAME
  • LanguagesEnglish
  • MaterialAdhesive Paper
  • Width (in)1
  • Height (in)1
  • Quantity250/roll
  • DescriptionGHS Label,Flame Over Circle Pictogram,1" x 1",adhesive paper,250/roll
Reviews
Q&A