Accuform LZH602EV5 GHS Label,Flame Over Circle Pictogram,1" x 1",adhesive poly,500/roll

Representative image only.

Accuform LZH602EV5 GHS Label,Flame Over Circle Pictogram,1" x 1",adhesive poly,500/roll

Accuform | Mfr: LZH602EV5| Item: 5071259

$70.28CAD / Roll of 500

$70.28CAD / Roll of 500
Product Description

More About this Item

GHS (Globally Harmonized System) is a worldwide program for classifying chemicals in the workplace. GHS recognizes nine hazard pictograms. The pictograms identify the hazard which helps users take proper precautions to minimize adverse effects.
GHS Flame Over Circle pictogram indicates the following potential hazard: oxidizers

Specifications
  • HeaderFLAME
  • LanguagesEnglish
  • MaterialAdhesive Poly
  • Width (in)1
  • Height (in)1
  • Quantity500/roll
  • DescriptionGHS Label,Flame Over Circle Pictogram,1" x 1",adhesive poly,500/roll
Reviews
Q&A