A&D Weighing HW-10KCEP Industrial Bench Scale, 20 lbs

Representative image only.

A&D Weighing HW-10KCEP Industrial Bench Scale, 20 lbs

A&D Weighing | Mfr: HW-10KCEP| Item: 1112700

Designed to weigh safely in hazardous environments

$4,014.99CAD / Each

for an additional
$304.26CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$4,014.99CAD / Each

Buy It Together

Current Item

A&D Weighing HW-10KCEP Industrial Bench Scale, 20 lbs
A&D Weighing HW-10KCEP Industrial Bench Scale, 20 lbs
1112700

$4,014.99CAD / Each
+ =
Total Price 4319.25 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Ten-digit keypad simplifies setting comparator limits
  • Stainless steel pan allows for wet weighing and easy cleanup
  • Built-in key lock prevents unwanted changes
  • Large LCD with white backlight enables viewing in dim light
  • IP65-rated base for both dust and water
  • IECEx, ATEX, and FM/FMc certified
  • Auto shut off saves battery life
  • 2 year warranty

More About this Item

The HW-CEP industrial bench scales are specially designed to weigh safely in potentially explosive atmospheres and are approved for use in FM Class 1, Division 1, groups C & D hazardous areas. Red, green and amber LED comparator lights allow you to increase productivity by allowing fast check weighing.

Other features include a 10-digit keypad for setting comparator limits, a key lock function to prevent unwanted setting changes, auto-tare function, accumulation function which displays sum total of separate weighing results, counting function, comparator function, external calibration, and low battery indicator. Weighing modes include parts counting and accumulation function for totaling weigh data. Weighing units include kg, lb, oz, and lb-oz.

The scales operate on 4 D batteries. No need to secure power from a safe area via a safety barrier.

Conformite EuropeenneIP 65Two year warranty
Specifications
  • Capacity (kg)10
  • Capacity (lbs)20
  • Repeatability (g)2
  • Linearity (g)2
  • Weighing Unitskg, lb, oz, lb-oz
  • Calibration TypeExternal
  • Stabilization Time (Sec)1.5
  • Auto ShutoffYes
  • Low Battery IndicatorYes
  • Battery4 D (not included)
  • Height (in)15 19/32
  • Width (in)9 51/64
  • Depth (in)17 19/64
  • Pan Length (in)9 51/64
  • Pan Width (in)9 51/64
  • Display Column Height (in)1
  • DescriptionIndustrial Bench Scale, 20 lbs
  • Warranty2 years
Reviews
Q&A