A&D Weighing GXA-25 External Fanless Ionizer (Static Eliminator)
Representative image only.
A&D Weighing GXA-25 External Fanless Ionizer (Static Eliminator)
A&D Weighing | Mfr: GXA-25| Item: 1110986
Communicate with your A&D balance using compatible output and relay kits
$1,583.89CAD / Each
$1,583.89CAD / Each
Product Description
Key Features
  • Eliminate static electricity from the weighing sample before the measurement, reducing weighing error
  • Compatible with A&D Apollo GF and GX series balances
  • 1 year warranty
More About this Item
An IR (touchless) switch is attached to the ionizer, and neutralization can be started without touching the ionizer. Power is supplied from the balance to allow the ionizer to be operated without using an AC
adapter.
1 Year Warranty
Specifications
  • DescriptionExternal Fanless Ionizer (Static Eliminator)