A&D Weighing EK-EP Instrinsically Safe Portable Balance, 3000 g x 0.1 g

Representative image only.

A&D Weighing EK-EP Instrinsically Safe Portable Balance, 3000 g x 0.1 g

A&D Weighing | Mfr: EK-3000AEP| Item: 1113304

Weigh safely in potentially explosive areas

$3,293.03CAD / Each

for an additional
$304.26CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$3,293.03CAD / Each

Buy It Together

Current Item

A&D Weighing EK-EP Instrinsically Safe Portable Balance, 3000 g x 0.1 g
A&D Weighing EK-EP Instrinsically Safe Portable Balance, 3000 g x 0.1 g
1113304

$3,293.03CAD / Each
+ + =
Total Price 4329.93 ADD 3 ITEMS TO CART
Product Description

Includes

AC power adapter.

Key Features

  • Intrinsically safe construction prevents spark generation
  • Compact, lightweight and portable—goes anywhere!
  • Durable 304 SS weighing pan is chemically resistant
  • Large backlit LCD is viewable in any lighting conditions
  • Comparator function provides fast accurate filling, packing, and sorting
  • Adjust response characteristics to compensate for drafts and vibrations
  • Use a cable lock with the built-in security ring to prevent theft
  • 2 year warranty

More About this Item

Featuring a compact and lightweight design and an easy-to-read LCD screen, these EK-EP Intrinsically Safe Weighing Scales are IECEx compliant, providing an effective weighing solution for a range of applications, including hazardous area weighing.

Highly portable, this IECEx scale can easily be taken on the go, making it a popular choice among the many IECEx weighing systems available. And because it’s an intrinsically safe weighing scale, you can trust that the top pan balance will provide precise weight measurement even in potentially explosive or hazardous environments and workplaces.

Two year warrantyFM Approved
Specifications
  • Capacity (g)3000
  • Readability (g)0.1
  • Repeatability (mg)0.1
  • Linearity (g)0.2
  • Weighing Unitsg, ct, oz, lb, mom, %, pcs (counting), N, dwt, gn, oz t, sg (specific gravity)
  • Calibration TypeExternal, with calibration mass
  • Battery4 AA (not included)
  • Height (in)2 1/8
  • Width (in)7 1/2
  • Depth (in)8 5/8
  • Pan Length (in)6 5/8
  • Pan Width (in)5 1/4
  • DescriptionInstrinsically Safe Portable Balance, 3000 g x 0.1 g
  • Warranty2 Years
Reviews
Q&A