3M H10A Earmuff, 30 dB NRR; 1/Pk
Representative image only.
3M H10A Earmuff, 30 dB NRR; 1/Pk
3M | Mfr: 3MRH10A| Item: 4849230
Dual shell minimizes resonance to give you maximum high-frequency attenuation.
$76.03CAD / Each
BUY MORE & SAVE
  • 1-9$76.03 CAD / Each
  • 10+$68.43 CAD / Each
$76.03CAD / Each
BUY MORE & SAVE
  • 1-9$76.03 CAD / Each
  • 10+$68.43 CAD / Each
Product Description
More About this Item
Dual shell design minimizes resonance for maximum high-frequence attenuation. Ear cushions are extra wide for secure fit and low surface pressure. Compliance:ANSI S3.19-1974
Specifications
  • NRR (dB)30
  • ComplianceANSI S3.19-1974
  • DescriptionEarmuff, 30 dB NRR; 1/Pk