3M FT-10 Respirator Qualitative Fit Test Kit, Sweet taste
Representative image only.
3M FT-10 Respirator Qualitative Fit Test Kit, Sweet taste
3M | Mfr: FT-10| Item: 8633230
Meets OSHA requirements
$773.53CAD / Each
$773.53CAD / Each
Buy It Together
Current Item
3M FT-10 Respirator Qualitative Fit Test Kit, Sweet taste
3M FT-10 Respirator Qualitative Fit Test Kit, Sweet taste
8633230
$773.53CAD / Each
+ =
Total Price 835.85 ADD 2 ITEMS TO CART
Product Description
Includes
sensitivity solution, fit test solution, two nebulizers, test hood, collar, and instruction booklet (English/French/Spanish/Portuguese).
Key Features
  • Easy to use
  • Provides a consistent environment for testing fit
More About this Item
These fit test kits test the face-to-respirator seal of particulate respirators and gas/vapor respirators with a particulate prefilter. Easy-to-use kits provide a consistent environment for testing fit, leading to reliable results meeting OSHA requirements.
Testing is easy: a sensitivity test is performed first by spraying a dilute version of the test solution into the hood without a respirator being worn to make sure the person can detect the taste. Then, while wearing a respirator, a sweet or bitter test solution is sprayed into the hood. If the wearer doesn't detect the taste of the test agent, the respirator fit is acceptable. If the taste is detected, a second test is performed after readjusting the respirator. If that test also fails, it may indicate the need for a different size or model of respirator.
Specifications
  • DescriptionRespirator Qualitative Fit Test Kit, Sweet taste