3M C-SKFL5 Folded Sorbent, Chemical Kit; 5-Gallon Capacity
Representative image only.
3M C-SKFL5 Folded Sorbent, Chemical Kit; 5-Gallon Capacity
3M | Mfr: C-SKFL5| Item: 4011700
Four sorbent configurations in one!
$123.47CAD / Each
BUY MORE & SAVE
  • 1-2$123.47 CAD / Each
  • 3+$111.12 CAD / Each
$123.47CAD / Each
BUY MORE & SAVE
  • 1-2$123.47 CAD / Each
  • 3+$111.12 CAD / Each
Product Description
More About this Item
Use these versatile folded sorbents as a boom, pillow, pad, or roll. Chemical and petroleum spill kits have a 5-gallon capacity, and come with a temporary disposal bag and tie. Use the maintenance sorbents for cleanup of non-aggressive liquids. Maintenance sorbents are perforated every 16", and come with a handy dispenser.
Specifications
  • Absorption5 gal.
  • Width (in)5
  • Length (in)26
  • DescriptionFolded Sorbent, Chemical Kit; 5-Gallon Capacity