3M 8233 Specialty Particulate Disposable Respirators, N100, Cool Flow valve
Representative image only.
3M 8233 Specialty Particulate Disposable Respirators, N100, Cool Flow valve
3M | Mfr: 8233| Item: 4011114
Industry standard for protection against solid and liquid particles
$563.57CAD / Box of 20
BUY MORE & SAVE
  • 1-9$563.57 CAD / Box of 20
  • 10+$507.21 CAD / Box of 20
$563.57CAD / Box of 20
BUY MORE & SAVE
  • 1-9$563.57 CAD / Box of 20
  • 10+$507.21 CAD / Box of 20
Product Description
Key Features
  • NIOSH approved
More About this Item
These lightweight respirator masks offer comfort and adjustable straps for a snug fit.
N100 Respirator offers maximum filter efficiency—use for grinding, and working with metals and arsenic. Not to be used with any oil aerosols.
Specifications
  • RatingN100
  • ComplianceNIOSH
  • ColorWhite
  • 3M Item Number8233
  • DescriptionSpecialty Particulate Disposable Respirators, N100, Cool Flow valve