3M 60926 PACK Filter cartridge, Multigas and vapor; P100
Representative image only.
3M 60926 PACK Filter cartridge, Multigas and vapor; P100
3M | Mfr: 60926 PACK| Item: 4011375
$74.70CAD / Pkg of 2
BUY MORE & SAVE
  • 1-29$74.70 CAD / Pkg of 2
  • 30+$63.50 CAD / Pkg of 2
$74.70CAD / Pkg of 2
BUY MORE & SAVE
  • 1-29$74.70 CAD / Pkg of 2
  • 30+$63.50 CAD / Pkg of 2
Buy It Together
Current Item
3M 60926 PACK Filter cartridge, Multigas and vapor; P100
3M 60926 PACK Filter cartridge, Multigas and vapor; P100
4011375
$74.70CAD / Pkg of 2
+
3M 6100 Half-mask respirators, Small
4011300
$37.28CAD / Each
=
Total Price 111.98 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
Replace your respirator cartridge according tot he demands of your work environment.
Specifications
  • RatingP100
  • NIOSH ApprovalCertain organic vapors, Chlorine, Hydrogen Chloride, Sulfur Dioxide, Chlorine Dioxide, Hydrogen Fluoride, Ammonia/Methylamine, Hydrogen Sulfide (escape only), Formaldehyde
  • ComplianceNIOSH
  • ColorGreen, Pink
  • DescriptionFilter cartridge, Multigas and vapor; P100