3M 50051138290080 Foam Earplugs, Uncorded; 200 Pairs/Box
Representative image only.
3M 50051138290080 Foam Earplugs, Uncorded; 200 Pairs/Box
3M | Mfr: 1100| Item: 8311100
Soft comfortable earplugs conform to a wide range of ear sizes
$94.00CAD / Box of 150
BUY MORE & SAVE
  • 1-9$94.00 CAD / Box of 150
  • 10+$84.60 CAD / Box of 150
$94.00CAD / Box of 150
BUY MORE & SAVE
  • 1-9$94.00 CAD / Box of 150
  • 10+$84.60 CAD / Box of 150
Product Description
More About this Item
Soft foam tapered earplugs conform to the unique shape of each ear for excellent hearing protection and comfort. The soft smooth foam seals the ear comfortably while resisting dirt for hygiene. Foam is made of hypo-allergenic material, and is a bright orange color for high visibility and ease of inspection. Plugs can be combined with earmuffs for additional protection. Packaged one pair per bag. Recommended for protection in moderate to high noise environments of up to 100 dBA TWA.
Compliance: ANSI S3.19-1974, CSA A(L)
Specifications
  • NRR (dB)29
  • ComplianceANSI S3.19-1974; CSA class A(L)
  • CordedNo
  • DescriptionFoam Earplugs, Uncorded; 200 Pairs/Box